Learn More
Medchemexpress LLC Trans-(1R,2R)-N-[(2S)-1-[4-(5-bromo-2-oxo-3H-benzimidazol-1-yl)piperidin-1-yl]propan-2-yl]-2-phenyl | 1246303-14-9 | 98.1% | 497.43 g·mol⁻¹ | C25H29BrN4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10129310MM 1ML
Encompass_Preferred
VU0359595 is a selective pharmacological inhibitor of phospholipase D1 (PLD1) supplied as a ready-to-use solution for research applications. It is offered as a 10 mM solution in DMSO and as a solid, providing flexibility for assay setup and compound handling. The material is reported at high purity and is intended for biochemical and cellular studies of PLD1-dependent signaling pathways.
  • Selective phospholipase D1 inhibitor
  • Ready-to-use 10 mM solution in DMSO
  • High purity, 98.06%
  • Suitable for biochemical and cellular assays
  • Also available as solid in multiple packaging sizes
Catalog No. 50-002-12651