Learn More
Medchemexpress LLC Trans-2,3,4-trimethoxycinnamic acid | 33130-03-9 | 96.5% | 238.24 | 10 G

Supplier: Medchemexpress LLC HYW00877010G
Trans-2,3,4-Trimethoxycinnamic acid is a phenylpropanoids product isolated from the barks of Chinese cinnamon, intended for research use only.
- Molecular weight of 238.24
- Molecular formula C12H14O5
- Appears as a white to off-white solid
- Store as powder at -20°C for 3 years, or 4°C for 2 years
- Store in solvent at -80°C for 6 months, or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.