Learn More
Medchemexpress LLC Trans-2,3,4-trimethoxycinnamic acid | 33130-03-9 | 96.5% | 238.24 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW008770100G
Encompass_Preferred
trans-2,3,4-Trimethoxycinnamic acid is a Phenylpropanoids product that can be isolated from the barks of Chinese cinnamon. It is for research use only and not sold to patients.
  • Solid appearance
  • White to off-white color
  • Initial source from plants Lauraceae Cinnamomum cassia (L.) D. Don
  • Ships at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-53722