Learn More
Medchemexpress LLC Trans-2,5-diethylpiperazine | 6189-24-8 | 142.25 | 250 MG

Supplier: Medchemexpress LLC 250MGHY24239A
trans-2,5-Diethylpiperazine is a drug intermediate for the synthesis of various active compounds. This product is for research use only and not sold to patients.
- Used as a drug intermediate
- Solid appearance
- Molecular formula C8H18N2
- Shipping at room temperature in continental US
- Storage at 4°C, protected from light
- Longer storage in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.