Learn More
Medchemexpress LLC Trans-2-hexenal | 6728-26-3 | 99.6% | 10 G
SDP
Supplier:  Medchemexpress LLC HY12842910G
Encompass_Preferred
trans-2-Hexenal ((E)-2-Hexenal) is a volatile compound widely present in fresh plants, vegetables, and fruits, known for its distinct leafy aroma. It exhibits antifungal activity and can also hinder the germination of soybean seeds and the growth of seedlings. Additionally, trans-2-Hexenal is utilized for determining low-molecular-weight carbonyl compounds that react with biological nucleophiles in biological samples.
  • Widely present in fresh plants, vegetables, and fruits
  • Known for its distinct leafy aroma
  • Exhibits antifungal activity
  • Can hinder the germination of soybean seeds and the growth of seedlings
  • Utilized for determining low-molecular-weight carbonyl compounds that react with biological nucleophiles in biological samples
Catalog No. 50-003-42589