Learn More
Medchemexpress LLC Trans-3-(3-pyridyl)acrylic acid | 19337-97-4 | MFCD00006410 | 99.9% | 149.15 g·mol⁻¹ | C8H7NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017463
Encompass_Preferred
trans-3-(3-pyridyl)acrylic acid is a trans-3-aryl acrylic acid used in chemical research and synthesis. It appears as a white to off-white solid (C8H7NO2, MW 149.15 g·mol⁻¹) and has reported antiviral activity against tobacco mosaic virus. The compound is used as a synthetic intermediate and building block in organic and coordination chemistry and is supplied at high purity for laboratory use.
  • White to off-white solid with high purity.
  • Molecular formula C8H7NO2; molecular weight 149.15 g·mol⁻¹.
  • Reported antiviral activity against tobacco mosaic virus (TMV).
  • Useful synthetic intermediate and building block for organic synthesis.
  • Supplied in multiple package sizes suitable for research use.
Catalog No. 50-003-02323