Learn More
Medchemexpress LLC Trans-3-(3-pyridyl)acrylic acid | 19337-97-4 | MFCD00006410 | 99.9% | 149.2 g/mol | C8H7NO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01746310MM/1ML
Encompass_Preferred
trans-3-(3-pyridyl)acrylic acid is a trans-3-aryl acrylic acid reported to inhibit tobacco mosaic virus (TMV). This listing is supplied as a 10 mM solution in DMSO (1 mL) for research use; the compound is reported at high purity and has documented storage recommendations for solvent and powder forms.
  • Ready-to-use 10 mM solution in DMSO, supplied in a 1 mL vial.
  • High reported purity suitable for screening and research applications.
  • Reported antiviral activity against tobacco mosaic virus in the literature.
  • Compact vial format reduces preparation time and solvent handling.
  • Stable when stored under recommended conditions for solvent and powder forms.
  • Includes safety and quality documentation such as SDS and COA.
Catalog No. 50-002-99424