Learn More
Medchemexpress LLC Trans-3-(5-methoxy-2-methyl-1H-indol-3-yl)-1-(4-pyridinyl)-2-propen-1-one | 1363421-46-8 | 99.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY119624100MG
Encompass_Preferred
MOMIPP is a small-molecule PIKfyve inhibitor that induces macropinocytosis and methuosis in cultured cells, and is reported to penetrate the blood-brain barrier. It is used as a research tool to study endosomal trafficking, macropinocytosis-related cell death, and PIKfyve-mediated signaling.
  • Inhibits PIKfyve and induces macropinocytosis leading to methuosis.
  • Penetrates the blood-brain barrier, enabling central nervous system studies.
  • Active in low micromolar ranges in cell-based assays.
  • Molecular formula C18H16N2O2; molecular weight 292.33 g·mol⁻¹.
  • Supplied with high purity (99.0%) and available in multiple pack sizes including 100 mg.
  • Stable as powder at -20°C for extended storage; store solutions at -80°C for long-term.
Catalog No. 50-002-33618