Learn More
Medchemexpress LLC Trans-3-indoleacrylic acid | 29953-71-7 | MFCD00005633 | 99.4% | 187.19 g/mol | C11H9NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW015273A50MG
Encompass_Preferred
Trans-3-indoleacrylic acid (CAS 29953-71-7) is a tryptophan metabolite used in biochemical research and reported to inhibit ferroptosis in cell-based studies. It is provided as a high-purity solid and as a ready-to-use DMSO solution for in vitro applications.
  • High purity suitable for biochemical and cell biology assays.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Relevant for ferroptosis and metabolic pathway research.
  • Molecular weight and formula provided for analytical reference.
  • Supplied in small research-friendly packaging sizes.
Catalog No. 50-003-02464