Learn More
Medchemexpress LLC Trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride | 15942-08-2 | MFCD26967902 | 426.57 g/mol | C14H19Br2ClN2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW13005810MG
Encompass_Preferred
trans-4-(6,8-dibromo-1,2-dihydroquinazolin-3(4H)-yl)cyclohexanol hydrochloride is a brominated dihydroquinazoline hydrochloride salt documented as an impurity of ambroxol hydrochloride. It is provided for analytical and reference use in pharmaceutical research, principally for impurity identification, characterization, and analytical method development. Consult the supplier's Certificate of Analysis for authoritative purity and storage information.
  • Used for impurity profiling and analytical method development.
  • Hydrochloride salt form for improved stability and handling.
  • Molecular formula C14H19Br2ClN2O; molecular weight 426.57 g/mol.
  • Suitable as a reference standard for characterization of related syntheses.
  • Certificate of Analysis available for purity and storage specifications.
Catalog No. 50-002-82975