Learn More
Medchemexpress LLC Trans-4-aminocrotonic acid | 38090-53-8 | MFCD00673818 | 99.3% | 101.10 g·mol⁻1 | C4H7NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100800100MG
Encompass_Preferred
TACA (trans-4-aminocrotonic acid) is a neuroscience research reagent used to probe GABAergic signaling; it functions as a receptor agonist and affects GABA uptake and metabolism, and is provided with analytical documentation for experimental use.
  • Potent agonist of GABAA and GABAC receptors.
  • Acts as a GABA uptake inhibitor and substrate for GABA-T.
  • High purity: 99.33%.
  • Molecular weight 101.10 g·mol⁻1 and formula C4H7NO2.
  • Available in solid and solution pack sizes including 100 MG.
  • Datasheet, COA, and SDS available for quality and safety information.
Catalog No. 50-002-11945