Learn More
Medchemexpress LLC Trans-4-carboxy-L-proline | 64769-66-0 | 98.0% | 159.14 | C6H9NO4 | 1 MG
Supplier: Medchemexpress LLC HY1008361MG
Trans-4-carboxy-L-proline is a selective glutamate transporter inhibitor provided as a high-purity research chemical intended for laboratory use. Supplied as a powder with defined storage conditions, it is used in neuropharmacology and transporter research where controlled dosing and stability are required.
- Selective glutamate transporter inhibitor for neuropharmacology studies.
- High purity (98.0%) suitable for research applications.
- Powder form for flexible preparation and dosing.
- Stable when stored at -20°C for up to 3 years.
- Available in small research-scale quantities (e.g., 1 mg).
- For research use only; not for human or clinical use.