Learn More
Medchemexpress LLC Trans-4-carboxy-L-proline | 64769-66-0 | 98.0% | 159.14 g/mol | C6H9NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1008365MG
Encompass_Preferred
trans-4-Carboxy-L-proline is an amino acid derivative used as a selective glutamate transporter inhibitor in neuroscience research. Supplied as a powder with manufacturer-specified purity and storage guidance, it is used to modulate glutamatergic signaling in in vitro and biochemical studies.
  • Selective glutamate transporter inhibitor for research applications.
  • High purity suitable for biochemical and cellular assays.
  • Powder form enables flexible preparation and dosing.
  • Documented storage conditions support multi-month stability.
  • Soluble in common laboratory solvents such as DMSO for stock solutions.
Catalog No. 50-002-11477