Learn More
AdipoGen trans-4-Hydroxy-L-proline (100 g)
SDP
Supplier:  AdipoGen CDXT0254G100
Encompass_Preferred
Chemical. CAS: 51-35-4. Formula: C5H9NO3. MW: 131.13. Trans-4-Hydroxy-L-proline is a non-essential amino acid and a natural constituent of animal structural proteins such as collagen and elastin. It is commonly used as a chiral building block in the organic synthesis of various compounds, such as neuroexcitatory kainoids and antifungal echinocandins. It has been found in many bacterial secondary metabolites, including echinocandins.
Catalog No. 50-205-4663