Learn More
Medchemexpress LLC HY-113974 10mg Medchemexpress, trans-AUCB CAS:885012-33-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11397410MG
Encompass_Preferred
Medchemexpress, HY-113974 10mg trans-AUCB CAS:885012-33-9 trans-AUCB (t-AUCB) is a potent, orally active and selective soluble epoxide hydrolase (sEH) inhibitor with IC50s of 1.3 nM, 8 nM, 8 nM for hsEH, mouse sEH and rat sEH, respectively. trans-AUCB has anti-glioma activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9279