Learn More
Medchemexpress LLC (+)-trans-chrysanthemic acid | 4638-92-0 | MFCD01941572 | 99.9% | 168.23 | C10H16O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW513778250MG
Encompass_Preferred
(+)-trans-Chrysanthemic acid is the trans enantiomer of chrysanthemic acid and a key intermediate used in the synthesis and study of pyrethroid insecticides. Supplied as a high-purity research chemical, it is intended for laboratory synthesis, insecticidal activity assays, and analytical method development.
  • High purity suitable for research and synthesis.
  • Functions as a pyrethroid component and synthetic intermediate.
  • Available in small gram-scale pack sizes for laboratory use.
  • Molecular formula C10H16O2; molecular weight 168.23.
  • Provided with supporting analytical documentation where available.
Catalog No. 50-002-84977