Learn More
Medchemexpress LLC Trans-Cinnamic acid | 140-10-3 | MFCD00004369 | 148.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN061010MM/1ML
Encompass_Preferred
Trans-Cinnamic acid is a natural antimicrobial agent. It exhibits minimal inhibitory concentration (MIC) of 250 μg/mL against the fish pathogen A. sobria, SY-AS1, and shows moderate inhibition against various other fish pathogens and isolates. Its effectiveness is enhanced when the pH of the culture media is not neutralized.
  • Antimicrobial activity against fish pathogens
  • Long-term stability when stored as solid or in solvent
  • Shipping at room temperature
Catalog No. 50-003-89086