Learn More
Medchemexpress LLC (-)-trans-Permethrin | 54774-47-9 | 99.8% | 10 MM 1 ML

Supplier: Medchemexpress LLC HYB0887B10MM/1ML
(-)-trans-Permethrin is a pyrethroid that provides satisfactory protection from black flies for cattle. It is for research use only.
- Pyrethroid that protects against black flies
- For research use only
- High purity of 99.76% (100.00% ee.)
- Available as a 10 mM solution in 1 mL of DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.