Learn More
AdipoGen trans-Piceatannol (25 mg)
SDP
Supplier:  AdipoGen CDXP0592M025
Encompass_Preferred
Chemical. CAS: 10083-24-6. Formula: C14H12O4. MW: 244.24. Piceatannol is a resveratrol analog with anticancer, anti-inflammatory, antioxidant, neuroprotective and cardioprotective properties. It is a spleen tyrosine kinase (Syk) inhibitor, however it exhibits antiproliferative activity by inhibiting a wide range of tyrosine and serine/threonine protein kinases, including Syk, p56lck, PKA, PKC, MLCK, CDPK, JNK and PI3K. It suppresses NF-kappa activation through inhibition of IkappaBalpha kinase and NLRP3 activation. It is an apoptosis and autophagy inducer, cell cycle inhibitor, activator of SIRT1 and also inhibits adipogenesis.
Catalog No. 50-205-4619