Learn More
Medchemexpress LLC HY-112301A 5mg Medchemexpress, trans-Pralsetinib CAS:2097132-93-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112301A5MG
Encompass_Preferred
Medchemexpress, HY-112301A 5mg trans-Pralsetinib CAS:2097132-93-7 trans-Pralsetinib (trans-BLU-667) is a rearranged during transfection (RET) inhibitor extracted from patent US20170121312A1, Compound Example 129. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0815