Learn More
Medchemexpress LLC Trans-R-138727 | 239466-74-1 | 99.0% | 349.42 g/mol | C18H20FNO3S | 5MG

Supplier: Medchemexpress LLC HY123669A5MG
trans-R-138727 is the trans stereoisomer of R-138727, the thiol-containing active metabolite of prasugrel. Supplied as a solid research reagent, it is used to study P2Y12 receptor-mediated platelet inhibition, pharmacology, and related biochemical assays.
- Trans stereoisomer of R-138727 and active metabolite of prasugrel.
- Irreversible P2Y12 receptor inhibitor used in platelet function research.
- High purity (≈99%) solid powder suitable for analytical work.
- Chemical formula C18H20FNO3S; molecular weight 349.4 g/mol.
- Recommended storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
- Available in small research sizes, including 5 mg vials.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.