Learn More
Medchemexpress LLC Trans-resveratrol | 501-36-0 | 100.0% | 228.24 g/mol | C14H12O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16561R10MG
Encompass_Preferred
Resveratrol (Standard) is an analytical reference standard of trans-resveratrol supplied as a high-purity solid for research and analytical applications. It is typically provided in 10 mg packages (CAS 501-36-0; formula C14H12O3; molecular weight 228.24 g/mol). See the SDS and Certificate of Analysis for storage, handling, and exact purity information.
  • High purity for accurate analytical results.
  • Intended for assay calibration and method validation.
  • Solid, white to off-white appearance for convenient handling.
  • Includes CAS number for unambiguous substance identification.
  • Chemical formula and molecular weight documented for reference.
  • Typical 10 mg package suitable for small-scale analytical use.
  • Refer to SDS and Certificate of Analysis for storage and safety details.
Catalog No. 50-002-89565