Learn More
American Radiolabeled Chemicals Inc TRANS TRANS FPP-3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 0324250UC
Encompass_Preferred
trans,trans Farnesyl pyrophosphate-1-3H triammonium salt 250uCi in 70% EtOH in 0.25 M NH4HCO3 under argon, 1 mCi/ml, S.A. 15-30 Ci/mmol, M.W. 433.4, Shipped in Dry Ice, (FPP)
Catalog No. 50-901-04370