Learn More
Medchemexpress LLC trans-trans-Muconic acid-d4 | 1955496-83-9 | 99.8% | 146.13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY113247S1MG
Encompass_Preferred
trans-trans-Muconic acid-d4 is the deuterium labeled trans-trans-Muconic acid. It is a urinary metabolite of benzene and has been used as a biomarker of exposure to benzene in humans. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37004