Learn More
Medchemexpress LLC trans-Zeatinriboside | 6025-53-2 | 99.8% | 351.36 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0111515MG
Encompass_Preferred
trans-Zeatinriboside is a type of cytokinin precursor that acts as a major long-distance signaling form in xylem vessels, regulating leaf size and meristem activity-related traits.
  • Cytokinin precursor
  • Major long-distance signaling form in xylem vessels
  • Regulates leaf size
  • Regulates meristem activity-related traits
  • For research use only
Catalog No. 50-004-52312