Learn More
Cayman Chemical trans211methylDodecenoic Aci
SDP
Supplier:  Cayman Chemical 100083211 MG
Encompass_Preferred
trans-.DELTA.2-11-methyl-Dodecenoic acid is the conformational isomer of cis-.DELTA.2-11-methyl-dodecenoic acid, a diffusible signal factor (DSF) in extracellular microbial and fungal communication systems.{13766}
Catalog No. 50-288-9888