Learn More
Medchemexpress LLC Transcription Factor EB Antibody (YA3266) | 50 UL

Supplier: Medchemexpress LLC HYP8352150UL
The Transcription Factor EB Antibody (YA3266) is a rabbit-derived antibody designed to react with human proteins. It is supplied as a liquid formulation suitable for various laboratory applications.
- Host: Rabbit
- Reactivity: Human
- Clonality: YA3266
- Molecular weight: Predicted band size 53 kDa; Observed band size 65-70 kDa
- Concentration: 0.3 mg/mL
- Non-conjugated
- Applications include WB, IHC-F, IHC-P, ICC/IF, and IP
- Formulated in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
- Store at -20°C for 1 year
- Avoid repeated freeze / thaw cycles
- Ships with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.