Learn More
Lonza Walkersville 4D-Nufe LV Reservoir Rack
Supplier:  Lonza Walkersville AAK3001
Encompass
The 4D-Nucleofector LV Reservoir Rack supports more convenient handling of 4D-Nucleofector LV Reservoirs Two racks are provided together with the 4D-Nucleofector LV Unit An additional rack might be required when working with the second input line for feeding the LV Nucleocuvette Cartridge
Catalog No. 50-004-96463