Learn More
Lonza Walkersville Prim Cel Opt 384-well Nufe Kit
Supplier:  Lonza Walkersville V5SP9001
Encompass
The Primary Cell Optimization 384-well Nucleofector Kit is suited for high-throughput optimization of transfection conditions for primary cell types lacking an Optimized Protocol for the 384-well HT Nucleofector System Complete Optimization Different conditions can easily be tested within one experiment using a 384-well Nucleocuvette Plate and our pmaxGFP Vector as positive control Our five Primary Cell Nucleofector Solutions P1 - P5 are tested in combination with a pre-selected set of programs plus controls A subsequent optional fine tuning of the Nucleofection conditions can be performed with the help of our scientific support team Cell Group-Specific Optimization using Basic Protocols In addition to a complete optimization described above we also provide a cell group-specific optimization protocol for some primary cell groups
Catalog No. 50-004-96432