Learn More
Lonza Walkersville Transfection Transfection Kit 1/PK
Supplier:  Lonza Walkersville VPL1004
Encompass
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfectiion of primary mouse or rat hepatocytes The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type
Catalog No. 50-004-96454