Learn More
Lonza Walkersville Cel Line Opt 384-well Nufe Kit
Supplier:  Lonza Walkersville V5SC9001
Encompass
The Cell Line Optimization 384-well Nucleofector Kit is suited for high-throughput optimization of transfection conditions for primary cell types lacking an Optimized Protocol for the 384-well HT Nucleofector System Different conditions can easily be tested within one experiment using a 384-well Nucleocuvette Plate and our pmaxGFP Vector as positive control Our three Cell Line Nucleofector Solutions SE SF and SG are tested in combination with a pre-selected set of programs plus controls A subsequent optional fine tuning of the Nucleofection conditions can be performed with the help of our Scientific Support Team Before doing an optimization for your cell type of interest please check out our knowledge database for availability of a ready-to-use Lonza Optimized Protocol or any existing data
Catalog No. 50-004-96388