Learn More
Lonza Walkersville Nufe Kit Prim Mamm Glial Cells
Supplier:  Lonza Walkersville VPI1006
Encompass
In addition to various cell type-specific kits for primary glial cells we also provide a cell group-specific Basic Nucleofector Kit This kit is suited to determine the optimal transfection conditions for any mammalian glial cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II) In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for glial cells The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types
Catalog No. 50-004-96364