Learn More
Sigma Aldrich Fine Chemicals Biosciences Transfer buffer powder for use with mPAGE(R) Bis-Tris gels pack of 10 ea (Each packets makes 1 liter of transfer buffer at 1X) |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MPTRB
Encompass
Transfer buffer powder for use with mPAGE(R) Bis-Tris gels pack of 10 ea (Each packets makes 1 liter of transfer buffer at 1X) |
Catalog No. NC2384552