Learn More
TWD Scientific LLC Transfer Pipette, Twdbiox, 3Ml, Disposable, Sterile, Polyethylene, Molded Graduations, Individually Wrapped, 2000/Case.
SDP
Supplier:  TWD Scientific LLC BX300138A11
Encompass_Preferred
3Ml Disposable Polyethylene Sterile General Purpose Transfer Pippets (Length: 160Mm | Capacity: 7.5Ml). Pippets Have Molded Graduations At Each 0.25Ml Up To 3Ml (22 Drops/Ml). Pippets Come Individually Wrapped. 500 Pipettes/Pack, 4 Packs/Case, Price/Case
Catalog No. 50-228-5029