Learn More
Sigma Aldrich Fine Chemicals Biosciences Transferrin HOLO huma500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SRP6512500MG
Encompass
Transferrin is a monomeric glycoprotein found in plasma at an average concentration of 250 mg/100ml. Transferrin (Tf) belongs to the family of bilobal glycoproteins which tightly bind ferric iron. Each of the homologous N- and C-lobes of the protein contains a single iron-binding site present in a deep cleft.
Catalog No. 50-290-6614