Learn More
Medchemexpress LLC Transferrin Mouse 100ug
SDP
Supplier:  Medchemexpress LLC HYP73581100UG
Encompass_Preferred
Transferrin is an important iron-binding transport protein that combines two Fe(3 ) ions with an anion usually bicarbonate Its main function is to transport iron from the site of absorption to the site of storage and utilization Transferrin Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Transferrin protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80011