Learn More
Alpco Ltd Mouse Transferrin ELISA
SDP
Supplier:  Alpco Ltd 41TRAMSE01
Encompass
Mouse Transferrin ELISA is for the quantitative determination of transferrin in mouse serum and plasma. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-288