Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peak Scientific Instruments Ltd Dual Tap Transformer for 195V - 219V
SDP

Supplier:  Peak Scientific Instruments Ltd 063210

Encompass

Dual Tap Transformer for 195V - 219V

Catalog No. NC1704633


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.