Learn More
Cusabio Technology LLC Human Transforming Growth factor B1TGF-B1 ELISA kit
Supplier:  Cusabio Technology LLC CSBE04725H
Encompass_Preferred
The product CSB-E04725h is a ready-to-use microwave, strip plate ELISA kit designed to quantitatively analyze the TGFB1 in different biological samples, including serum, plasma, and cell culture supernates. This TGFB1 ELISA kit specifically recognizes the human TGFB1 in the sample. It adopts the Sandwich ELISA technique in conjunction with an enzyme colorimetric reaction to indicate the presence of human TGFB1 and quantitatively measures the level of TGFB1 in the samples. The intensity of the colored product is positively related to the amount of human TGFB1 in the sample.
Catalog No. NC3654118