Learn More
Medchemexpress LLC Transient receptor potential canonical 5 (TRPC5) activator | 896684-04-1 | 99.8% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12820510MG
Encompass_Preferred
BTD is a selective transient receptor potential canonical 5 (TRPC5) activator. It can be utilized for research related to neurological diseases.
  • Selective activator of transient receptor potential canonical 5 (TRPC5).
  • Activates TRPC5 with EC50 values of 1.4 μM (fluorometric microwell Ca2+ influx analyses) and 1.3 μM (in whole cell patch clamp experiments).
  • Activates TRPM8-expressing cells with an EC50 value of 20.6 μM.
  • Capable of activating heteromeric channel complexes that include TRPC5 and its closely related channels.
Catalog No. 50-003-40601