Learn More
XpressBio TRANSIL Membrane Affinity Kit (8 vials)
SDP
Supplier:  XpressBio TMP04010008
Encompass_Preferred
The Sovicell TRANSIL Membrane Affinity kit enables researchers to measure the interaction of drugs with a membrane. This in vitro assay is designed to measure the distribution coefficient of test compounds between phosphatidylcholine membranes and an aqueous buffer. We refer to this distribution coefficient as membrane affinity.
Catalog No. NC2956611