Learn More
Medchemexpress LLC Transketolase-IN-2 | 2757552-21-7 | C18H9F3N2O3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15160050MG
Encompass_Preferred
Transketolase-IN-2 (compound 4w) is a potent Transketolase inhibitor. It demonstrates strong inhibition of *Digitaria sanguinalis* and *Amaranthus retroflexus* (over 90% at 200 mg/L and approximately 80% at 100 mg/L). It can be used in studies of weed control.
  • Potent transketolase inhibitor
  • Strong inhibitory effects against *Digitaria sanguinalis* (92% root inhibition, 90% stem inhibition at 200 mg/L)
  • Strong inhibitory effects against *Amaranthus retroflexus* (91% root inhibition, 90% stem inhibition at 200 mg/L)
  • Useful in studies of weed control
Catalog No. 50-003-72063