Learn More
Medchemexpress LLC Transketolase-IN-4 | 419547-73-2 | C19H14ClN3O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15309010MM/1ML
Encompass_Preferred
Transketolase-IN-4 is a potent transketolase inhibitor with an IC50 of 3.9 μM. It inhibits the proliferation of tumor cells such as SW620, LS174T, and MIA PaCa-2. This compound is also a possible Mycobacterium tuberculosis DXS inhibitor, with an IC50 value of 114.1 μM.
  • Potent transketolase inhibitor with an IC50 of 3.9 μM
  • Inhibits proliferation of tumor cells including SW620, LS174T, and MIA PaCa-2
  • Possible Mycobacterium tuberculosis DXS inhibitor with an IC50 of 114.1 μM
  • White to off-white solid appearance
Catalog No. 50-003-72238