Learn More
GA International Transparent Permanent Cryogenic & Autoclave-Resistant Laser Labels 4" x 0.625" / 101.6mm x 15.9mm Rectangle Transparent From -196°C to +150°C (-321°F to +302°F)
Supplier:  GA International DFLT77
Encompass_Preferred
Cryogenic thermoplastic transparent laser labels for long-term storage in liquid nitrogen and deep-freeze harsh environments for biomedical research and clinical laboratories. The labels are also resistant to steam autoclave sterilization and boiling water. These transparent cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.
Catalog No. 50-005-71783