Learn More
Medchemexpress LLC Transthyretin/TTR M 1mg
SDP
Supplier:  Medchemexpress LLC HYP735801MG
Encompass_Preferred
The transthyretin/TTR protein is a thyroid hormone binder that may transport thyroxine from the blood to the brain It exists as a homotetramer forming a dimer of dimers with a ligand-regulated central channel suggested to play a role in thyroxine binding and transport Transthyretin/TTR Protein Mouse (HEK293 His) is the recombinant mouse-derived Transthyretin/TTR protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-80010