Learn More
Medchemexpress LLC Tranylcypromine hydrochloride | 1986-47-6 | 99.7% | 169.66 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY17447A10MM/1ML
Encompass_Preferred
Tranylcypromine hydrochloride (SKF 385 hydrochloride) is an irreversible inhibitor of lysine-specific demethylase 1 (LSD1/BHC110) and monoamine oxidase (MAO). It inhibits LSD1, MAO A, and MAO B with IC50s of 20.7, 2.3, and 0.95 μM, respectively. This product can be used for the research of depression.
  • Irreversible inhibitor of LSD1/BHC110 and monoamine oxidase (MAO)
  • Inhibits LSD1, MAO A, and MAO B with specific IC50 values
  • Can be used for the research of depression
Catalog No. 50-003-86591