Learn More
CPC Scientific H-Ser-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific PARP018B
Encompass
This peptide is the fragment 42-47 of the thrombin receptor, which acts as a thrombin receptor activator. It is effective in causing tyrosine phosphorylation, inhibition of cAMP formation, and an increase in cytosolic Ca2+.SEQUENCE: H-Ser-Phe-Leu-Leu-Arg-Asn-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: SFLLRNMOLECULAR FORMULA: C34H56N10O9MOLECULAR WEIGHT: 748.88STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141136-83-6]SYNONYMS: Thrombin Receptor Activator Peptide 6, Thrombin Receptor (1-6) (human), PAR-1 (1-6) (human), Proteinase Activated Receptor 1 (1-6) (human), SFLLRN, Coagulation Factor II Receptor (1-6) (human)RESEARCH AREA: HematologyREFERENCES:L.F.Brass et al., J. Biol. Chem., 269, 2943 (1994)R.Kaufmann et al., NeuroReport, 9, 709 (1998)B.A.Olson et al., Pharmacology, 58, 200 (1999)H.Andersen et al., Proc. Natl. Acad. Sci. USA, 96, 11189 (1999)R.Kaufmann et al., J. Neurooncol., 42, 131 (1999)C.-C.Wu et al., Br. J. Pharmacol., 130, 1289 (2000)H.
Catalog No. 50-211-0126