Learn More
Wilmad Labglass TRAP ROT-EVP 100ML 24/40 19/22
Supplier:  Wilmad Labglass LG6881102
Encompass_Preferred
Trap, Rotary Evaporator, Drain Back. Size: 100mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 19/22 Inner. Base of inner tube has two 3mm openings to allow drain-back of foam condensate to the evaporator flask.
Catalog No. 50-803-689