Learn More
Wilmad Labglass TRAP VAC 2PC 38X225MM COMP
Supplier:  Wilmad Labglass LG11026102
Encompass_Preferred
Vacuum Trap, Two-Piece, with Hose Connectors, Complete. Body O.D.: 38mm; Body Length: 225mm; Joint: [ST] 34/45; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture
Catalog No. 50-807-213
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.