Learn More
Wilmad Labglass TRAP VACUUM 1-PC 35X250MM HC
Supplier:  Wilmad Labglass LG11013104
Encompass_Preferred
Vacuum Trap, One-Piece. With Hose Connectors. Body O.D.: 35mm; Body Length: 250mm; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture.
Catalog No. 50-803-995